Imprimer
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit
Fournisseur: AAT BIOQUEST
22415
AATB22415EA 460 EUR
AATB22415
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit
Tests Tests immunologiques
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit provides a novel one-step fluorescence assay for the determination of gram sign in living bacteria.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for fluorescent microscopy
Attention: For research use only.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR