Imprimer
MycoLight™ Bacterial Viability Assay Kit
Fournisseur: AAT BIOQUEST
22400
AATB22400EA 460 EUR
AATB22400
MycoLight™ Bacterial Viability Assay Kit
Tests Tests immunologiques
Mycolight™ Bacterial Viability Assay Kit provides two-colour fluorescence assay of bacterial viability in both gram-positive and negative bacterial cell.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
Attention: For research use only.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR