Imprimer

XFD568 acid fluorescent probe

Fournisseur: AAT BIOQUEST
Attention

1792
AATB1792EA 439 EUR
AATB1792
XFD568 acid fluorescent probe
XFD568 acid

XFD568 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 568 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 807.74 g/mol
ADR: 5 II

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR