Imprimer
Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in Water*
Fournisseur: AAT BIOQUEST
17710
AATB17710EA 138 EUR
AATB17710
Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in Water*
Colorants Colorants pour gels d'electrophorèse
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Attention: For research use only.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR