Imprimer

Z-IETD-FMK

Fournisseur: AAT BIOQUEST
Attention

13305
AATB13305EA 143 EUR
AATB13305
Z-IETD-FMK
Z-IETD-FMK

Z-IETD-FMK is a cell-permeable, irreversible caspase-8 inhibitor.


  • Caspase inhibitor
  • Enhanced stability
  • Increased cell permeability

Formule: C₃₀H₄₃FN₄O₁₁
MW: 654.68 g/mol
Numéro MDL: MFCD03490491
Numéro CAS:  210344-98-2
ADR: 5 II

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR