Imprimer

Luciferase imaging_luminescent

Fournisseur: AAT BIOQUEST
Attention

12500
AATB12500EA 152 EUR
AATB12500
Luciferase imaging_luminescent
Luciferase

This enzyme is a luciferase expressed from a cloned gene from the North American firefly (Photinus pyralis) that provides the reliability and dependability needed for performing research or producing kits using bioluminescence reagents to detect ATP or luciferin substrates.


  • Reliable
  • Significantly decreased variability

MW: 60 kDa
Numéro CAS: 61970-00-1
ADR: 5 II

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR