Imprimer

Red 780 Maleimide, mFluor™ fluorescent probe

Fournisseur: AAT BIOQUEST
Attention

1192
AATB1192EA 439 EUR
AATB1192
Red 780 Maleimide, mFluor™ fluorescent probe
Red 780 Maleimide

mFluor™ Red 780 dyes are an excellent alternative to APC-Alexa Fluor® 750 tandems since they have the spectral properties equivalent to those of APC-Alexa Fluor® 750 conjugates.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 1168.28 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=546610a2fc103049f47e406989f845fa91b0d0e7&vid=3fad88978b8e610e0e1e202005e9124ec5d4ea6b&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR