Imprimer
VWR®, Withdrawal System for 30 L Stainless Steel Barrels, Horizontal
Fournisseur: VWR Chemicals
99901.030EA 575 EUR
99901.030
VWR®, Withdrawal System for 30 L Stainless Steel Barrels, Horizontal
Pompe Pompes pour conteneur/transfert
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 30 L stainless steel barrels.
  • Withdrawal system for 30 L barrels (stainless steel)
  • Perfluorelastomer o-rings
  • Includes 1 dip tube for 30 L barrels
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 30 L returnable stainless steel barrels.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR