Imprimer
VWR® Hitachi ChromAssist, data station
Fournisseur: VWR Collection
906-0123EA 1990 EUR
906-0123 906-0124 HITA903-0374 HITA903-0375
VWR® Hitachi ChromAssist, data station
Logiciels Chromatography Software
The newly developed ChromAssist Data Station combines the proven intuitive structure of the Hitachi software family with an innovative new design. As a chromatography data system (CDS), ChromAssist contains all the necessary functions for controlling VWR® Hitachi HPLC and UHPLC instruments, including method creation, sample table, data integration and reporting.
  • New user-friendly design
  • Integrated training videos and help functions
  • Simple creation of methods, sample tables
  • Innovative report editor
  • Advanced GLP functionality
Additionally, someMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR