Imprimer

Tétrahydrofuranne ≥99.0% (par GC, corrigé de la teneur en eau) stabilisé, AR® ACS, Macron Fine Chemicals™

Fournisseur: Avantor Performance Materials
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

8498-26EA 0 EUR
8498-26
Tétrahydrofuranne ≥99.0% (par GC, corrigé de la teneur en eau) stabilisé, AR® ACS, Macron Fine Chemicals™
Tétrahydrofuranne
Formule: C₄H₈O
MW: 72.11 g/mol
Point d'ébullition: 66 °C (1013 hPa)
Point de fusion: –108.5 °C
Densité: 0.888 g/cm³ (20 °C)
Point d'éclair: –21.5 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00005356
Numéro CAS: 109-99-9
EINECS: 203-726-8
ONU: 2056
ADR: 3 II
ATTEINDRE: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (clear, colorless liquid) Passes Test
ACS - Assay (C₄H₈O) (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤20
ACS - Peroxide (as H₂O₂)(at time of packaging) ≤0.015 %
ACS - Residue after Evaporation ≤0.03 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Contains 0.025% Butylated Hydroxytoluene as a stabilizer.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR