Imprimer
Blot module, miniVE™
Fournisseur: Cytiva
80-6418-96
80-6418-96EA 637 EUR
80-6418-96
Blot module, miniVE™
Appareils de transfert
Perform 1-D and 2-D electrophoresis and electrotransfer in a single compact unit.
  • Complete semi-wet blotting in 45 minutes with only 300 ml buffer
  • Choose precast gels from a variety of manufacturers
  • Perform two second-dimension separations using 7 cm IPG strips in 15 hours
Cast, run, and blot mini-gels in four streamlined components: Gel module, blot module, lower buffer chamber, safety lid.
Two mini-gels or four mini-blots at once, using 100×105 mm gel cassettes for increased separation distances.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR