Imprimer
Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Fournisseur: G Bioscienes
786-789
786-789EA 895 EUR
786-789
Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Protéines purification Protein Purification Resins
Immobilised Pepstatin is for the purification of cathepsins, pepsin, bacterial aspartic proteases, HIV proteases and other molecules that bind pepstatin.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR