Imprimer
Nucleic acid and protein destain, Silver D-Stain™
Fournisseur: G Bioscienes
786-199
786-199EA 478 EUR
786-199
Nucleic acid and protein destain, Silver D-Stain™
Réactifs d'électrophorèse Réactifs détachants
The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stain™ eliminates the need to repeat the electrophoresis.
Suitable for reducing the staining of silver stained gels and revealing over-stained bands.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR