Imprimer
Phosphate buffered saline (PBS), 10X
Fournisseur: G Bioscienes
786-027
786-027EA 99.7 EUR
786-027
Phosphate buffered saline (PBS), 10X
Tampons Tampons d'électrophorèse Tampons de lavage
PBS 10X solution is used as a wash buffers for Western blotting, ELISA and other applications.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR