Imprimer
Laveurs de microplaques, Wellwash™ Versa
Fournisseur: THERMO LABSYSTEMS LIFE SCIENCE
5165010 5165050
736-0420EA 8770 EUR
736-0420 736-3009
Laveurs de microplaques, Wellwash™ Versa
Laveurs de microplaques
Wash 96- or 384-well plates securely with the Thermo Scientific™ Wellwash™ Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications. Designed for high performance and versatility, enjoy the ease of use and convenience provided through a graphical interface, local language version and USB port.
  • Easy and convenient to use
  • Large color display and keypad for easy, convenient use
  • Clear display shows everything on one screen
  • User interface is logic and intuitive, allowing quick protocol development
  • Easy-to-use software
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR