Imprimer
Microplate readers, multi-mode, SpectraMax® Mini
Fournisseur: Molecular Devices
SMAXMINIAFL SMAXMINIAF SMAXMINIAL 5089096 5089097 5089098 5089099 5089100 5089102 5089103 5089104 5089334
735-0396EA 0 EUR
735-0396 735-0395 735-0397 MLDV5089096 MLDV5089097 MLDV5089098 MLDV5089099 MLDV5089100 MLDV5089102 MLDV5089103 MLDV5089104 MLDV5089334
Microplate readers, multi-mode, SpectraMax® Mini
Lecteurs de microplaques
The SpectraMax® Mini multi-mode microplate reader offers a budget friendly solution that gives you exactly what you need when you need it. With three modes of detection for UV/Vis absorbance, fluorescence and luminescence, the SpectraMax® Mini reader is user-upgradeable - from two to three modes, providing researchers with a versatile instrument that does not compromise on sensitivity or performance.
  • Industry leading software - enjoy an extensive library of protocols, advanced reader control, and easy data management and analysis with our 'best in class'
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR