Imprimer
VWR®, Ensembles de flacons, PETG, stériles, à usage unique
Fournisseur: VWR Collection
734-2844EA 940 EUR
734-2844 734-2845 734-2846 734-2850 734-2851 734-2894
VWR®, Ensembles de flacons, PETG, stériles, à usage unique
Flacons Flacons pour milieux de culture
Sterile PETG containers, with weldable tubings, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They come in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR