Imprimer
24 clear not treated reservoir, sterile, HTS Transwell®
Fournisseur: Corning
3395
734-1558EA 336.1 EUR
734-1558
24 clear not treated reservoir, sterile, HTS Transwell®
Microplaques Microplaques HTS
For use as reservoir tray with HTS Transwell permeable supports.
  • Not treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR