Imprimer
Plaques de prélèvement et d'analyse, Whatman UNIPLATE™
Fournisseur: Whatman products (Cytiva)
7701-5110
734-1217EA 421 EUR
734-1217
Plaques de prélèvement et d'analyse, Whatman UNIPLATE™
Microplaques Microplaques deepwell
PP, with clear PS lid. The Whatman™ UNIPLATE collection and analysis microplates accomodate 24 well plates, have round bottom wells.
  • Chemical compatibility: Available in chemically resistant polymers capable of withstanding low temperatures for long-term storage. Opaque plates prevent optical cross talk in light emitting assays
  • Guaranteed for use with robotic handlers and centrifuge carriers
Certifications: UNIPLATE microplates conform to the ANSI/SBS microplate standard and fit most microplate readers and automated plate handling devices.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR