Imprimer
Chamber slides, Corning® BioCoat™ Poly-D-lysine/laminin
Fournisseur: Corning
354688
734-1072EA 481 EUR
734-1072
Chamber slides, Corning® BioCoat™ Poly-D-lysine/laminin
Lames Chambres de culture
These culture slides have a uniform application of poly-D-lysine/laminin.
  • Tested to promote neurite outgrowth
  • NG108 rat glioma/mouse neuroblastoma cells
  • PS vessel lid together with removal tool
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR