Imprimer

Electroporation system, twin-wave, Gemini X² and SC

Fournisseur: BTX
45-2044 45-2040 45-2041 45-2042 45-2043
732-3775EA 20810 EUR
732-3775 732-3771 732-3772 732-3773 732-3774
Electroporation system, twin-wave, Gemini X² and SC
Systèmes d'électroporation Électroporateurs
Gemini twin-wave electroporators are flexible systems allowing both square wave and exponential decay wave electroporation in a single unit. These waveform combinations enable researchers to easily and efficiently electroporate eukaryotic and prokaryotic cells in cuvettes or plates with one easy to use set-up. Ideal for all the electroporation needs, including CRISPR, in vivo, in vitro, in ovo and more.

  • Universal electroporation – transfects cells in vitro, in vivo, in ovo and adherent forms
  • Multiple pulsing capabilities in both square and exponential
More Product Information
Commander maintenant

SPÉCIFICATIONS

L×P×H 286×216×324 mm 286×216×324 mm 286×216×324 mm
Modèle Gemini X2 HT Gemini X2 Gemini SC
Environmentally Preferable
Interfaces Simple user interface Simple user interface Simple user interface
Temps de charge <7 s <7 s <7 s
Plage de tension mode "High Voltage" / résolution 5 V 505 - 3000 V 505 - 3000 V 505 - 3000 V
Plage de tension mode "High Voltage" / résolution 10 V 510 - 3000 V
Plage de tension mode "Low Voltage" / résolution 1 V 5 - 500 V 5 - 500 V
Salves d'impulsions 1 - 10 pulses per sample (up to 120 total) 1 - 99 pulses per sample 1 - 10 pulses per sample (low voltage mode);
1 - 2 pulses per sample (high voltage mode)
Pulse interval 0,1 - 10 s; 5 - 30 s 0,1 - 10 s; 5 - 30 s 0,1 - 10 s
Plage de longueur d'impulsion, mode HV/résolution 1 µs 10 - 600 μs 10 - 600 μs
Plage de longueur d'impulsion, mode LV/résolution 1 µs 10 - 999 μs 10 - 999 μs
Plage de longueur d'impulsion, mode LV/résolution 1 ms 1 - 999 ms 1 - 999 ms 10 - 100 ms
Capacité en mode "High Voltage" 85 µF 85 µF 85 µF
Capacité en mode "Low Voltage" 3775 µF 3775 µF 3775 µF
Affichage Touch screen Touch screen Touch screen
Poids 7,3 kg 7,3 kg 7,3 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR