Imprimer
Plaques d'électroporation
Fournisseur: BTX
45-0466 45-0467 45-0450 45-0452
732-0480EA 104 EUR
732-0480 732-0481 732-0476 732-0477
Plaques d'électroporation
Plaques d'électroporation
Sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR