Imprimer
VWR®, UV Transilluminator, Benchtop
Fournisseur: VWR Collection
730-1488EA 1350 EUR
730-1488
VWR®, UV Transilluminator, Benchtop
Transilluminateurs Transilluminateurs UV
The VWR® benchtop transilluminator for stand alone imaging systems is designed for ease of use.
  • High intensity output that equals or betters performance of models with higher wattage tubes
  • Includes a UV protective lid
  • Available in one size - 20×20 cm
  • Available with mid wave UV (302 nm) excitation wavelength
  • Choice of filter sizes
  • Safety cut-off (when in darkroom)
Even Illumination- across the filter surface due to special reflector/diffuser to eliminate tube patterns from images- this is essential when images are to be quantified with accuracy.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR