Imprimer
Sliding microtome, HM 450
Fournisseur: Epredia
910020
720-2708EA 21060 EUR
720-2708
Sliding microtome, HM 450
Microtomes et groupes de microtomes Microtomes
This HM 450 includes an automatic fine sectioning and trimming features with selectable specimen retraction.
  • Integrated and removable waste tray
  • Specimen fine orientation on X/Y axes
  • Memory function and graphical display in two different sizes
Suitable for section paraffin and frozen sections in biological, botanical, and materials science applications.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR