Imprimer
Multi-Parameter Tester, CombiCheck 80
Fournisseur: WTW
250489
713-4103EA 170 EUR
713-4103
Multi-Parameter Tester, CombiCheck 80
Kits de tests environnementaux
CombiCheck solutions are ready-to-use multi-parameter standards, Each package contains a standard solution and an addition solution. Both solutions can be used directly without dilution for quality assurance.
  • The standard solution is used to check the accuracy of the results for the complete system: procedure - analytical method - reagents - photometer
  • The addition solution is used to check sample-dependent influences by measuring the recovery rate and to determine the most suitable sample preparation method
The maximum number of determinations which can be made with a CombiCheck standard solution depends on the test set used. With the addition solution 280 determinations are always possible.
Parameters can be checked with their concentrations as follows:
COD: 1,500 mg/L COD
Nitrate: 25,0 mg/L NO₃-N
Phosphate: 15,0 mg/L PO₄-P
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR