Imprimer
Yeast extract, Veggie™
Fournisseur: Merck Millipore (Novagen)
71279-3
71279-3EA 266 EUR
71279-3
Yeast extract, Veggie™
Milieux pour microbiologie Additifs et suppléments de microbiologie
Veggie yeast extract is a certified animal-free media component.
  • Ensures proper growth and maintenance of bacterial cells
  • Ideal for applications requiring minimal use of animal-derived materials
Used as a direct replacement for traditional yeast extract in bacterial growth media.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR