Imprimer
Coliform swab
Fournisseur: HYSERVE
1000101
710-1479EA 237 EUR
710-1479
Coliform swab
Écouvillons Écouvillons d'échantillonnage
Easy to carry early warning test for contamination of surfaces with coliform bacteria. Based on a certified method, the Coliform Swab provides a simple method for the qualitative detection of these bacteria - with a clear colour change from purple to yellow in the presence of coliform bacteria.
  • Simple colour change shows contamination, no colour change indicates the corresponding contamination
  • Individually packed straightening test, which can be carried out without additional reagents
  • Results can be read within 18 hours
  • Determines the number of germs from 1
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR