Imprimer
Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ3700
Fournisseur: Thermo Orion
AQ3700
705-0732EA 2070 EUR
705-0732
Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ3700
Colorimètres Colorimètres multiparamètres
Measure common colorimetric water and wastewater parameters using the Orion™ AQUAfast™ AQ3700 colorimeter with 90 preprogrammed methods, automatic selection of 6 LED wavelengths and timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank.
  • Wide range of disinfectants in various formats to broaden our offering to a variety of industries
  • Large graphic display with step-by-step user instructions
  • Automatic countdown to measurement
  • Consistent results from shift to shift
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR