Imprimer

Supelco® Organic Reference Standard, δ-Tocotrienol

Fournisseur: Merck

69745-100MG
69745-100MGEA 1300 EUR
69745-100MG
Supelco® Organic Reference Standard, δ-Tocotrienol
Étalons Étalons organiques

Commander maintenant

SPÉCIFICATIONS

Synonyme(s) (R)-δ-Tocotrienol; 8-Methyltocotrienol; [R-(E,E)]-3,4-Dihydro-2,8-dimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol
Environmentally Preferable
M-Clarity quality level MQ100
N ° CAS. 25612-59-3
MDL No. MFCD11045309
Molecular weight 396,61
Conditions de transport DICE
Conditions de stockage –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR