Imprimer
Orion™ 9502BNWP Carbon Dioxide Electrode
Fournisseur: Thermo Orion
9502BNWP
662-1591EA 1110 EUR
662-1591
Orion™ 9502BNWP Carbon Dioxide Electrode
Électrodes Électrodes ISE
This electrode measures carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately.
  • Gas-sensing combination electrode
  • With waterproof BNC connector
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR