Imprimer
Electrode Adapter Cables
Fournisseur: Thermo Orion
91CBNC 91USRF
662-1535EA 113 EUR
662-1535 ORIO91USRF
Electrode Adapter Cables
Câbles
These cables can be used to convert an electrode with one connector to the required connector of another meter.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins or just a stripped lead inputs.
Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR