Imprimer
Histology bath, pfm 1000
Fournisseur: PFM Medical GmbH
041010
632-0060EA 4000 EUR
632-0060
Histology bath, pfm 1000
Bains Bains tissulaires
The housing of the pfm Waterbath 1000 is made from one complete block of aluminium. The tank is black anodised aluminium. The light anodised surface is easy to clean, the bath is ideal for stretching paraffin sections in the histology laboratory. It can be used in routine practice, research and industry.
  • Clear digital touch-screen with electronic temperature control
  • Water level control via sensor
  • Over-heating protection to avoid overheating and damage even when the bath is empty
  • Water temperature can be adjusted quickly and precisiely
  • Power supply is
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR