Imprimer
Flexacam C5 and I5 Microscope Cameras
Fournisseur: LEICA MICROSYSTEMS
12730537 12730535 12730536
630-3528EA 1790 EUR
630-3528 630-3526 630-3529
Flexacam C5 and I5 Microscope Cameras
Caméras
With Flexacam microscope cameras, you can see the fine sample details you need to see with precise colors and high resolution.
  • Flexacam c5 microscope camera for C-mount installation on a stereo or compound microscope
  • Flexacam i5 microscope camera for installation between the optics carrier and binocular tube of a stereo or compound microscope
  • Reveal fine details of your sample and capture images in high resolution thanks to the 12-megapixel CMOS sensor
  • Differentiate details quickly and clearly due to the fast auto exposure, even when switching between
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR