Imprimer
Viscosimètres, ViscoSystem® AVS® 370
Fournisseur: SI Analytics
1056509 1056515 1056516 1056517 1056518
616-0028EA 14610 EUR
616-0028 616-0034 SCOI1056516 616-0067 SCOI1056518
Viscosimètres, ViscoSystem® AVS® 370
Viscosimètres Viscosimètres capillaires automatiques
ViscoSystem AVC 370 combines a high degree of measurement accuracy and reproducibility with modularity.
  • Measure by suction (samples with high viscosity) and under pressure (liquids with low boiling points)
  • Modular design: Up to four ViscoPump II modules
  • Multiple measurement station controllable via a PC and multitasking software
  • WinVisco 370 software for two rinsing methods
  • Virtually all standard capillary viscometers can be used
The software allows control of up to two complete AVS 370 systems or a total of eight ViscoPump II modules. The results canMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR