Imprimer
SRK® Surfaces and Equipment Microbial Testing Devices
Fournisseur: Copan
902C 904C 905C 908C T2905C 922C 926C T2905CS01 T2906C T2906CS01 939CS01 940C 941C 929C 947C 958C 961C 906C.CR 961CS01 952C 948C 906C 949C 9U002S 9U004S 920C 9U017S 9U011S01 9U012S 924C 9U007N 9U005N
710-0472EA 27.3 EUR
710-0472 710-0474 710-0475 710-0431 710-0480 710-0477 710-0478 710-0481 710-0482 710-0483 612-2678 612-2679 612-2680 710-0932 710-0933 COPA958C COPA961C 710-1976 COPA961CS01 710-1862 COPA948C 710-1975 COPA949C 710-1977 COPA9U004S COPA920C 710-2232 710-2231 COPA9U012S COPA924C COPA9U007N COPA9U005N
SRK® Surfaces and Equipment Microbial Testing Devices
Écouvillons Écouvillons d'échantillonnage
Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.
  • Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR