Imprimer
Analyseurs d'humidité, professionnel, série MB 90 et MB 120
Fournisseur: OHAUS
30246943 30246944 30241169 30303303 30303304
611-3579EA 3350 EUR
611-3579 611-3580 611-3571 611-3746 611-3747
Analyseurs d'humidité, professionnel, série MB 90 et MB 120
Analyseurs Analyseurs d'humidité
Moisture analysers simplify complex moisture analysis applications. Designed to increase precision and efficiency in the laboratory, they feature a precisely controlled halogen heating system which gives fast, accurate results.
  • Colour touch screen display with intuitive icon-based navigation
  • Easy, tool-free cleaning for convenient maintenance
  • Quickly connects to a PC or flash drive
  • Precisely controlled, uniform, fast halogen heating technology provides repeatable results
  • Define drying parameters and store up to 100 methods with MB 120
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR