Imprimer

Acide nitrique, Finyte® Microelectronic, J.T.Baker®

Fournisseur: Avantor

Synonyms: Nitric acid solution

5801-13EA 260 EUR
5801-13
Acide nitrique, Finyte® Microelectronic, J.T.Baker®
Acide nitrique
Formule: HNO₃
MW: 63.01 g/mol
Point d'ébullition: 87
Point de fusion: -42
Densité: 1.51
Numéro MDL: MFCD00011349
Numéro CAS: 7697-37-2
ONU: 2031
ADR: 8 II
Merck Index: 13,06610

Commander maintenant

Résultats des tests de spécification

For Microelectronic Use 69.0 - 70.0 %
Assay (HNO₃) 69.0 - 70.0 %
Appearance Passes Test
Color (APHA) ≤5
Chloride (Cl) ≤0.05 ppm
Fluoride (F), For Information Only = 0.05 ppm
Phosphate (PO₄) ≤0.05 ppm
Sulfate (SO₄) ≤0.2 ppm
Trace Impurities - Aluminum (Al) ≤5.0 ppb
Arsenic and Antimony (as As) ≤3.0 ppb
Trace Impurities - Barium (Ba) ≤5.0 ppb
Trace Impurities - Beryllium (Be) ≤5.0 ppb
Trace Impurities - Bismuth (Bi) ≤5.0 ppb
Trace Impurities - Boron (B) ≤5.0 ppb
Trace Impurities - Cadmium (Cd) ≤5.0 ppb
Trace Impurities - Calcium (Ca) ≤10.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤5.0 ppb
Trace Impurities - Copper (Cu) ≤5.0 ppb
Trace Impurities - Gallium (Ga) ≤5.0 ppb
Trace Impurities - Germanium (Ge) ≤5.0 ppb
Trace Impurities - Gold (Au) ≤5.0 ppb
Trace Impurities - Indium (In) ≤20.0 ppb
Trace Impurities - Iron (Fe) ≤5.0 ppb
Trace Impurities - Lead (Pb) ≤5.0 ppb
Trace Impurities - Lithium (Li) ≤5.0 ppb
Trace Impurities - Magnesium (Mg) ≤5.0 ppb
Trace Impurities - Manganese (Mn) ≤5.0 ppb
Trace Impurities - Molybdenum (Mo) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤5.0 ppb
Trace Impurities - Niobium (Nb) ≤5.0 ppb
Trace Impurities - Platinum (Pt) ≤20.0 ppb
Trace Impurities - Potassium (K) ≤5.0 ppb
Trace Impurities - Silicon (Si) ≤5.0 ppb
Trace Impurities - Silver (Ag) ≤5.0 ppb
Trace Impurities - Sodium (Na) ≤5.0 ppb
Trace Impurities - Strontium (Sr) ≤5.0 ppb
Trace Impurities - Tantalum (Ta) ≤5.0 ppb
Trace Impurities - Thallium (Tl) ≤5.0 ppb
Trace Impurities - Tin (Sn) ≤5.0 ppb
Trace Impurities - Titanium (Ti) ≤5.0 ppb
Trace Impurities - Vanadium (V) ≤5.0 ppb
Trace Impurities - Zinc (Zn) ≤5.0 ppb
Trace Impurities - Zirconium (Zr) ≤5.0 ppb
Particle Count - 0.5 µm and greater ≤50 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR