Imprimer
Nitrogen generator, Solaris
Fournisseur: PEAK SCIENTIFIC
65-0001
564-0277EA 7000 EUR
564-0277
Nitrogen generator, Solaris
Générateurs de gaz Générateurs d'azote
This nitrogen generator has been engineered and designed as a gas delivery solution for ELSD, Compact Mass Spectrometer instruments and TLD readers which can reduce downtime and increase workflow efficiencies.
  • Space saving design
  • Variable outlet flow, pressure and purity
  • Optional Solaris air compressor stackable module
  • Colour indicative LED lighting for easy status awareness
The generator can be placed on a bench top and paired with an additional air compressor unit to provide air supply for labs without an in-house air supply or those who wish toMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR