Imprimer

Ammonium formiate 99 Anhydrous basis eluent additive, LC-MS, Fluka™

Fournisseur: Honeywell Chemicals
Attention

Synonyms: Formic acid ammonium salt

55674-50G
55674-50GEA 223 EUR
55674-50G
Ammonium formiate 99 Anhydrous basis eluent additive, LC-MS, Fluka™
Ammonium formiate
Formule: HCO₂NH₄
MW: 63.06 g/mol
Point de fusion: 116 °C
Densité: 1.266 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00013103
Numéro CAS: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Commander maintenant

Résultats des tests de spécification

Assay (calculated on the anhydrous basis) 99.0 - 101.0 %
Water (Karl Fischer) Max. 2 %
Sulfated ash Max. 0.1 %
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 0.1 ppm
Barium (Ba) Max. 1 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 5 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 1 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 5 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 1 ppm
Manganese (Mn) Max. 1 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 1 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Suitability for the LC-MS complying
Appearance of the solution complying
Solution 1 M in H₂O complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR