Imprimer
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Fournisseur: VWR Collection
548-1995AEA 28.1 EUR
548-1995A 548-1996A 548-1997A 548-1998A 548-1999A 548-2000A 548-1511A 548-1443A 548-1445A 548-1444A 548-1447A 548-1448A 548-1446A 548-1512A 548-1513A
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Boîtes Boîtes pour flacons
A broad variety of PP storage boxes to securely hold a wide range of vial sizes and bottles. Perfect for space-saving storage in the laboratory or in fridges.
  • Transparent lid
  • Chemically resistant
  • Stackable and robust
  • Temperature resistant from –80 to 100 °C
  • Easy sample identification (alphanumeric coding) for boxes with 81 cavities
Transperent lids prevents condensation on the closures and thus avoid contamination in the refrigerator.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR