Imprimer
Acrylic Imhoff cone rack, Nalgene®
Fournisseur: Thermo Fisher Scientific
1001-0010
532-0100EA 251 EUR
532-0100
Acrylic Imhoff cone rack, Nalgene®
Supports de statif
Securely hold three Imhoff cones with the durable and transparent Thermo Scientific™ Nalgene™ Acrylic Imhoff Cone Rack, featuring a wide base for stability on the lab bench.
  • Transparent; easily see level of contents
  • Can also be used to hold 500 and 1000 ml Nalgene separatory funnels
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR