Imprimer
Savant™ SpeedVac™ SPD140P1 Medium Capacity Vacuum Concentrator
Fournisseur: THERMO SAVANT
SPD140P1-230
531-1430EA 28350 EUR
531-1430
Savant™ SpeedVac™ SPD140P1 Medium Capacity Vacuum Concentrator
Concentrateurs
Savant™ SpeedVac™ medium capacity vacuum concentrators evapourate strong acids, bases, and aggressive organic solvents. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
  • Equipped with radiant lamps within the chamber
  • Heated lid for processing high boiling points (DMSO, DMF) and independent timers for both heat and run
  • Audible alarm to alert user when operation is finished
  • Built-in vacuum gauge and vacuum display
  • Wider vapour port and tubing for enhanced vapour throughput,
More Product Information
Limited Time Sale
Certains articles ci-dessous sont en promotion
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
L×P×H 33×46×36 cm
Poids 26 kg
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR