Imprimer
VWR®, Centrifuge tubes, ASTM
Fournisseur: VWR Collection
525-1568EA 30.6 EUR
525-1568 525-1569
VWR®, Centrifuge tubes, ASTM
Tubes Tubes à centrifuger
Borosilicate glass 3.3, used for the determination of water and sediments in crude oils according to test method ASTM D 96.
  • High chemical and thermal resistance
  • High contrast blue enamel graduations
  • Withstand forces up to 700 ×g
Design, graduations and tolerances meet ASTM (American Society for Testing Materials) specifications.
Certifications:
ASTM E-982
Information de livraison: Delivered without cap.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR