Imprimer
Entonnoirs de Büchner
Fournisseur: KARTELL
437 438 439 440 441 442 443 445
519-2361EA 20.2 EUR
519-2361 519-2362 519-2363 511-0008 519-2364 519-2365 519-2366 519-2367
Entonnoirs de Büchner
Entonnoirs Entonnoirs pour la filtration Entonnoirs Büchner
PP, white.
  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR