Imprimer
Analyseurs de boissons Microcheck® II
Fournisseur: Cytiva (Formerly Pall Lab)
4761 4763 4764 4761ME
516-0201EA 188 EUR
516-0201 516-0203 516-0204 516-1227
Analyseurs de boissons Microcheck® II
Entonnoirs Entonnoirs filtrants avec membrane
MicroCheck™ II beverage monitor, is an easy-to-use disposable filter funnel designed to meet the needs for microbial analysis of beverages.
  • Membrane diameter: 47 mm
  • Diameter: 6,1 cm
  • Membrane size: 0,45 µm
Polypropylene construction eliminates compatibility or extractables issues. Ergonomically designed units allow for removal of funnel cylinder with a squeeze. The base and cover convert to a petri dish for culturing the membrane in place, or the membrane can be cultured on a separate petri dish. Unit fits directly onto a standard manifold, or aMore Product Information
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Dimensions 61Ø×76H mm
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR