Imprimer
Moisture test paper, Whatman™
Fournisseur: Whatman products (Cytiva)
5401-090E
515-2877EA 70.3 EUR
515-2877
Moisture test paper, Whatman™
Filtres Filtres de fibres de verre
These borosilicate glass papers are designed for use when drying samples during moisture assessment.
Max. recommended temperature: 550 °C
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR