Imprimer
Membrane filters, cellulose nitrate, sterile, Whatman™
Fournisseur: Whatman products (Cytiva)
10401170
514-2752EA 301 EUR
514-2752
Membrane filters, cellulose nitrate, sterile, Whatman™
Filtres Filtres à membrane
Recommended for microbiological culturing and sample preparation, these cellulose nitrate membrane filters offer very narrow pore size distribution and low levels of extractables.
  • Sterile membrane filters eliminate the risk of contamination that could impact analysis
  • High strength and flexibility
The cellulose nitrate sterile membrane filters are well suited for separation applications involving particles up to 12 µm. After sample preparation, filtration residue is almost completely retained by the plain white membrane surface, supporting clearMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR