Imprimer
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Fournisseur: Thermo Fisher Scientific
124-0045
513-1275EA 67.78 EUR
513-1275
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Filtres Unités de filtration sous vide
Rapid-Flow™ disposable filter units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination.
  • Color coded according to membrane
  • Fastest flow rate and lowest clogging of comparable alternatives
  • 0.1 µm PES available for mycoplasma protection
  • 0.2 µm PES devices are stem cell and mouse embryonic assay (MEA) tested
Ideal for the filtration of cell culture media, serum, additives and buffers.
Non-pyrogenic and endotoxin free - Rapid-Flow filter units
More Product Information
Limited Time Sale
Certains articles ci-dessous sont en promotion
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Certification environnementale ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR