Imprimer

5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin 95% Dye, Sigma-Aldrich®

Fournisseur: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: 4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl)tetrakis-Phenol, 5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine

MQ100

477567-1G 477567-250MG
477567-1GEA 268 EUR
477567-1G 477567-250MG
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin 95% Dye, Sigma-Aldrich®
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin

It may be used to prepare porphyrin based matrices for the detection of water soluble vitamins by matrix-assisted laser desorption/ionization (MALDI) mass spectrometry. It may be used to prepare 5,10,15,20-tetrakis (4-polyethyleneoxy)phenyl)) porphyrin. Nanoporous silica inserted with 2H-OHTPP imparts gas sensitivity, and it could be useful to detect NO2 by surface plasmon resonance.

Formule: C₄₄H₃₀N₄O₄
MW: 678.75 g/mol
Point de fusion: >300 °C (lit.)                      
Température de stockage: Room Temperature
Numéro MDL: MFCD00191501
Numéro CAS: 51094-17-8
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba3ae55b28a653bef39c65d4af71493727c911fc&vid=b17a0d64a380e1a4c83b1320650cdd72c507dae4&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Appearance (Colour)
Dark Violet or Purple
Conforms to Requirements
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength
C=0.001 G/L Methanol
417 - 423 nm
Extinction Coefficient ≥285000
Carbon 73.1 - 82.5%
Nitrogen 7.7 - 8.7%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR