Imprimer
Magnetic stirrers, TWISTER
Fournisseur: IKA
20027389 20027010 20027388 20027390 20112621
442-1328EA 1208 EUR
442-1328 442-1326 442-1327 442-1329 442-1334
Magnetic stirrers, TWISTER
Agitateurs Agitateurs magnétiques
TWISTER is a universal and compact mixing device with magnetic drive. In the default version, TWISTER is a powerful single-position magnetic stirrer for mixing up to 5 litres at a maximum speed of 3000 min⁻¹ and a viscosity of up to 500 mPa·s. By connecting additional TWISTER stirring units (available as accessories, IKAA20112621), TWISTER can be expanded to a multi-position magnetic stirrer with up to 30 stirring positions. The stirring positions can be controlled either separately or simultaneously (leader-follower principle).
  • Touch operation
  • Powerful mixing, ideal for mixing small
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR